Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin Epithelial Mouse anti-Bovine, Chicken, Human, Mouse, Monkey, Rat, Rabbit, Clone: AE3, MilliporeSigma™

Catalog No. MAB1611MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1611MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1611MI Supplier MilliporeSigma Supplier No. MAB1611
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Keratin Epithelial Antibody, clone AE3 is an antibody against Keratin Epithelial for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Keratin Epithelial
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone AE3
Conjugate Unconjugated
Formulation Liquid in PBS. Contains 0.1% sodium azide.
Gene Accession No. NM_002273.2
Gene Symbols KRT8, KO, K8, CYK8, Keratin-8, CK-8, Cytokeratin-8, K2C8, CK8, CARD2
Host Species Mouse
Immunogen Human epidermal cells
Quantity 500 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Monkey, Mouse, Rabbit, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.