Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin K3/K76 Mouse anti-Bovine, Human, Mouse, Rabbit, Clone: AE5, MilliporeSigma™

Catalog No. CBL218MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL218MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL218MI Supplier MilliporeSigma Supplier No. CBL218
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Keratin K3/K76 Antibody, clone AE5 is an antibody against Keratin K3/K76 for use in western blot, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Keratin K3/K76
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone AE5
Conjugate Unconjugated
Formulation The monoclonal was purified by ion exchange chromatography is presented in phosphate buffered saline (pH 7.2) containing 0.09% sodium azide.
Gene Accession No. NM_057088.2
Gene Symbols KRT3, CK3, K3, CK-3, Cytokeratin-3, Keratin-3
Host Species Mouse
Immunogen Rabbit corneal epithelial keratin
Purification Method Ion exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rabbit
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.