Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki-67 Mouse anti-Human, Unlabeled, Clone: 35, BD

Catalog No. BDB610969 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610969 150 μg
BDB610968 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610969 Supplier BD Biosciences Supplier No. 610969
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cellular proliferation is a complex multi-faceted process, central to biological events ranging from embryonic development to wound healing. Although proliferation is tightly controlled, dysregulation often results in tumorigenesis. The regulatory process involves the expression of many cell-cycle-associated proteins that are subject to cycle-dependent modification. Such protein expression may be monitored to assess the proliferative capacity of cells, especially tumorigenic cells.The Ki-67 antigen is a nuclear protein expressed exclusively in proliferating cells during all active parts of the cell cycle. However, it is absent in quiescent cells and during DNA repair. The distribution of Ki-67 changes during different stages of the cell cycle. During G1, it is localized to the perinuclear region, but is primarily found in the nuclear matrix in later phases. During mitosis, it associates with the condensed chromosomes. The nuclear localization of Ki-67 and strict association with the cell cycle indicate its importance in the regulation of cell division. Therefore, Ki-67 has become an important marker of proliferating cells, and may also be a marker for disitinct nuclear matrix compartments.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ki-67
Applications Western Blot
Classification Monoclonal
Clone 35
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Ki-67 aa. 1547-1742
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.