Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki-67, Mouse, Clone: B56, BV711, BD

Catalog No. BDB563755 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563755 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563755 Supplier BD Biosciences Supplier No. 563755
Add to Cart
Add to Cart

Host Species: Mouse
Clone: B56
Isotype: IgG1, κ
Species Reactivity: Hu, Ms, Po
Immunogen: Human Ki-67

Flow Cytometry (Intracellular Staining)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ki-67
Classification Monoclonal
Clone B56
Host Species Mouse
Immunogen Human Ki-67
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Pig
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.