Learn More
Description
Host Species: Mouse
Clone: B56
Isotype: IgG1, κ
Species Reactivity: Hu, Ms, Po
Immunogen: Human Ki-67
Flow Cytometry (Intracellular Staining)
Specifications
Specifications
| Antigen | Ki-67 |
| Classification | Monoclonal |
| Clone | B56 |
| Host Species | Mouse |
| Immunogen | Human Ki-67 |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human, Mouse, Pig |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.