Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki-67 Mouse, FITC, Clone: B56, BD

Catalog No. BDB556026 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556026 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556026 Supplier BD Biosciences Supplier No. 556026
Add to Cart
Add to Cart

Includes

FITC Mouse Anti-Human Ki-67, FITC Mouse IgG1, κ Isotype Control

Mouse Monoclonal Antibody

Recognizes Ki-67, a nuclear cell proliferation-associated antigen expressed in all active stages of the cell cycle. Ki-67 is revealed as a double band (345 and 395kDa) in western blot analysis of proliferating cells. B56 was developed using an immunogen composed of the immunodominant epitope of the Ki-67 protein. Antibodies B56 and MIB 1 react with this immunogen. Flow cytometric analysis reveals that the binding of B56-PE can be blocked by MIB 1 purified antibody.

Host Species: Mouse
Clone: B56 , MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human Ki-67

Flow Cytometry (Intracellular Staining)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ki-67
Applications Flow Cytometry
Classification Monoclonal
Clone B56
Conjugate FITC
Host Species Mouse
Immunogen Human Ki-67
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.