Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki67 Mouse anti-Human, APC, Clone: Ki-S5, MilliporeSigma™Antigen HostSpecies anti-Target, Conjugate, Clone: clone

Catalog No. FCMAB103AP Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB103AP 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB103AP Supplier MilliporeSigma Supplier No. FCMAB103AP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Ki67 using this Milli-Mark™ Anti-Ki67 Antibody, clone Ki-S5 APC conjugate validated for use in flow cytometry, immunocytochemistry, immunohistochemistry, flow cytometry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ki67
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone Ki-S5
Conjugate APC
Formulation Purified mouse monoclonal IgG1 conjugated to Allophycocyanin (APC) in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NM_002417.3
Gene Symbols MKI67, KIA, Ki-67
Host Species Mouse
Immunogen Nuclear protein preparation from the human cell line U937.50
Purification Method Protein A purfied
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.