Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KIAA1191, Clone: 2F7, Abnova™

Catalog No. 89300339 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-339 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-339 Supplier Abnova Corporation Supplier No. H00057179M01
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen KIAA1191
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant KIAA1191.
Formulation PBS with no preservative; pH 7.4
Gene KIAA1191
Gene Accession No. NM_020444.2
Gene Alias FLJ21022/p60MONOX
Gene Symbols KIAA1191
Host Species Mouse
Immunogen KIAA1191 (NP_065177.2, 1 a.a. ∼ 305 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57179
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.