Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1614, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant KIAA1614.

Supplier:  Abnova Corporation PAB22122

Catalog No. 89-123-829


Only null left
Add to Cart

Description

Description

Sequence: DVEVENEVKEGRGHTPEGTLFLREDAKPPDLELKRVSLGPQWQPGPGLGSHQPHPLDSRTPCRTAYATTAPMTPESSGPG
Specifications

Specifications

KIAA1614
Polyclonal
Rabbit polyclonal antibody raised against recombinant KIAA1614.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
RP11-46A10.3
Rabbit
Antigen affinity purification
RUO
57710
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user.
KIAA1614
KIAA1614
Recombinant protein corresponding to amino acids of human KIAA1614.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only