Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF1A Mouse anti-Mouse, Unlabeled, Clone: 16, BD

Catalog No. BDB612094 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612094 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612094 Supplier BD Biosciences Supplier No. 612094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ability of the kinesin superfamily of motor proteins to hydrolyze ATP as they move progressively along microtubules is important for organelle transport and cell division. Kinesins are grouped according to the location of the motor domain in the N-terminal, middle, or C-terminal region of the protein. A family of N-terminal motor domain kinesin proteins includes KIF1A/1B, KIF3A/3B, KRP85/95, and Klp68d/64. KIF1A and KIF1B are 93% homologous in their N-terminal motor domains, however KIF1A contains a C-terminal PH domain. KIF1A and KIF1B are expressed in neurons where KIF1A is involved in fast anterograde axon transport of synaptic vesicles, and KIF1B is involved in anterograde axon transport of mitochondria. KIF1A associates with organelles that contain synaptotagmin, synaptophysin, and Rab3A. Disruption of the KIF1A gene in mice causes decreases in synaptic vesicle density, neuronal degeneration, and deficits in motor and sensory function. Thus, KIF1A is a critical brain motor protein involved with axonal transport of synaptic vesicle precursors.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF1A
Applications Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse KIF1A aa. 902-1015
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.