Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-KIF1B Clone: 10F3.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABC309
Description
Specifically detects KIF1B Clone: 10F3.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
Kinesin-like protein KIF1B (UniProt O60333; also known as Kinesin superfamily protein KIF1B, Klp) is encoded by the KIF1B (also known as KLP, CMT2, CMT2A, CMT2A1, HMSNII, KIAA0591, KIAA1448, NBLST1) gene (Gene ID 23095) in human. KIF1B is a member of the kinesin 3 family of proteins. The KIF1B gene encodes four alternatively spliced isoforms, among which isoform 2 and isoform 3 are also known as the alpha and beta isoforms. The alpha and beta isoforms share a common N-terminal motor domain but contain different C-terminal cargo domains. The alpha and beta isoforms are motor proteins implicated in anterograde transport of mitochondria and synaptic vesicle precursors, respectively. In addition, the beta isoform is required for developmental apoptosis induction by nerve growth factor (NGF) deprivation among neural crest progenitors during sympathetic neuron development. The beta isoform is reported to interact with RNA helicase A (DHX9), resulting in DHX9 nuclear accumulation and upregulated transcription of the proapoptotic XIAP-associated factor 1 (XAF1). KIF1B gene is located in a chromosomal region frequently deleted in neural crest-derived tumors, including neuroblastomas with loss of KIF1Bβ expression and impaired DHX9 nuclear localization.
Specifications
KIF1B | |
Monoclonal | |
Unconjugated | |
O60333 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Expected to react with spliced isoforms 1 and 2 (isoform Beta), but not isoform 3 (isoform Alpha). Reactivity toward isoform 4 is highly likely, but not yet confirmed. | |
-20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot | |
10F3.1 | |
Mouse monoclonal IgG2aκ tissue culture supernatant without preservatives. | |
KIF1B, KIAA0591, KIAA1448 | |
GST-tagged recombinant protein corresponding to the C-terminal region of human KIF1B. | |
100 μL | |
Apoptosis, Cancer | |
NP_055889 | |
Human, Mouse, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction