Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF3A Mouse, Unlabeled, Clone: 28, BD

Catalog No. BDB611508 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611508 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611508 Supplier BD Biosciences Supplier No. 611508
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ability of the kinesin superfamily of motor proteins to hydrolyze ATP as they move progressively along microtubules is important for organelle transport and cell division. Kinesins are grouped according to the location of the motor domain in the N-terminal, middle, or C-terminal region of the protein. A family of N-terminal motor domain kinesin proteins includes KIF3A/3B, KRP85/95, and Klp68d/64. These proteins form heterodimeric two-headed motors that interact with kinesin associated protein (KAP). The KIF3A/3B/KAP3 heterotrimeric motor has plus-end directed microtubule sliding activity. This heterotrimeric motor is expressed ubiquitously and is used for anterograde transport of membranous organelles. KIF3A- and KIF3B-deficient mice have defects in the formation of the cilia located in the embryonic node. In addition, KIF3A localizes to the basal body of the connecting cilium axoneme and to vesicles at the pre-synaptic ribbon in photoreceptor cells. Thus, KIF3 motors are involved in microtubule-based formation and movement of cilia and the transport of synaptic vesicles.

Host Species: Mouse
Clone: 28
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human KIF3A aa. 563-671
Formula Weight [Chemical]: 80/85kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF3A
Applications Western Blot
Classification Monoclonal
Clone 28
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human KIF3A aa. 563-671
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.