Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF4A, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA131786

Catalog No. PIPA131786


Description

Description

This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.

PA1-31786 detects KIF4A from human samples.PA1-31786 has been successfully used in immunofluorescence, Western blot applications.The PA1-31786 immunogen is: Peptide with sequence SFFSGCSPIEEEAH, from C Terminus of the protein sequence according to NP_036442 (amino acids 1219-1232 of Human KIF4A).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KIF4A
Polyclonal
Unconjugated
O95239, P33174
KIF4, KIF4A
Peptide with sequence SFFSGCSPIEEEAH, from C Terminus of the protein sequence according to NP_036442 (amino acids 1219-1232 of Human KIF4A).
100 μg
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Western Blot
0.5mg/mL
KIF4A
FLJ12530, FLJ12655, FLJ14204, FLJ20631, HSA271784, KIF4, KIF4-G1
Goat
IgG fraction
RUO
16571, 24137
Store at -20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.