Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kindlin-3 Rat anti-Human, Clone: 181A, MilliporeSigma™

Catalog No. MABT187MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT187MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT187MI Supplier MilliporeSigma Supplier No. MABT187
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Kindlin-3 Antibody, clone 181A is an antibody against Kindlin-3 for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kindlin-3
Applications Western Blot
Classification Monoclonal
Clone 181A
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_113659
Gene Symbols FERMT3, KIND3, MIG2B, URP2
Host Species Rat
Immunogen His-tagged recombinant protein corresponding to human Kindlin-3.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.