Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin Mouse anti-Chicken, Mammals, Squid, Sea Urchin, Human, Mouse, Rat, Clone: H2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1614
Description
Detect Kinesin using this Anti-Kinesin Antibody, heavy chain, a.a.420-445, clone H2 validated for use in ELISA, immunocytochemistry, immunohistochemistry, immunoprecipitation, FUNC, RIA and western blotting.
Specifications
Kinesin | |
Monoclonal | |
Unconjugated | |
NP_004512 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG2b |
ELISA, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
H2 | |
Purified mouse monoclonal IgG2b in buffer containing 0.02MPhosphate buffer, 0.25MNaCl with 0.1% sodium azide. | |
KIF5B, UKHC, KINH, KNS1, KNS | |
Bovine brain kinesin. | |
100 μg | |
Chicken, Human, Mouse, Rat, Sea urchin, Squid | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction