Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin Mouse anti-Mammals, Clone: L2, MilliporeSigma™

Catalog No. MAB1617MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1617MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1617MI Supplier MilliporeSigma Supplier No. MAB1617
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Kinesin Antibody, light chain, clone L2 is an antibody against Kinesin for use in ELISA, immunoprecipitation, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone L2
Conjugate Unconjugated
Formulation Liquid in 0.02MPhosphate buffer, 0.25MNaCl with 0.1% sodium azide.
Gene Accession No. NM_005552.4; NM_182923.3
Gene Symbols KLC1, hKLC1R, KNS2, KLC, KNS2A, hKLC1S, hKLC1P, hKLC1N, MGC15245, hKLC1J, hKLC1G, hKLC1B
Host Species Mouse
Immunogen Bovine brain kinesin.
Quantity 100 μg
Primary or Secondary Primary
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.