Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kip1 (p27) Rabbit anti-Human, Rat, Rabbit, Polyclonal, MilliporeSigma™

Catalog No. 06445MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-445-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-445-MI Supplier MilliporeSigma Supplier No. 06445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Kip1 (p27) Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Kip1 (p27) and has been validated in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kip1 (p27)
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
Gene Symbols CDKN1B; p27Kip1; P27KIP1; MEN1B; MEN4; CDKN4; KIP1
Host Species Rabbit
Immunogen An 18 amino acid synthetic peptide ([C]GSVEQTPKKPGLRRRQT) linked to KLH was used to immunize rabbits
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004064
Target Species Human, Rabbit, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.