Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR-NKAT2 Mouse anti-Human, FITC, Clone: DX27, BD

Catalog No. BDB556070 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556070 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556070 Supplier BD Biosciences Supplier No. 556070
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with KIR-NKAT2, a member of the killer-inhibitory receptor (KIR) family. The KIR family consists of transmembrane glycoproteins of the Ig superfamily expressed on NK cells and a subset of T cells which are involved in recognition of MHC class I molecules on target cells and inhibit cytotoxicity. KIR-NKAT2 antigen is involved in the recognition of HLA-C molecules (-Cw3 and related alleles).

Host Species: Mouse
Clone: DX27
Isotype: IgG2a κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR-NKAT2
Applications Flow Cytometry
Classification Monoclonal
Clone DX27
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.