Learn More
anti-KIR2DL4/CD158d, APC, Clone: 181703, R&D Systems™
Description
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | KIR2DL4/CD158d |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 181703 |
| Conjugate | APC |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | Q99706 |
| Gene Alias | 103AS, 15.212, CD158 antigen-like family member D, CD158d, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor |
| Gene Symbols | KIR2DL4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.