Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KIR2DL4/CD158d, Fluorescein, Clone: 181703, R&D Systems™

Specifications

Antigen KIR2DL4/CD158d
Clone 181703
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB2238F025
View Documents
R&D Systems
FAB2238F025
25 Tests N/A N/A N/A
FAB2238F100
View Documents
R&D Systems
FAB2238F100
100 Tests N/A N/A N/A
Description

Description

Applications:

Flow Cytometry

Specifications

Specifications

KIR2DL4/CD158d
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
103AS, 15.212, CD158 antigen-like family member D, CD158d, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor
KIR2DL4
IgG2a
Protein A or G purified from hybridoma culture supernatant
Stains human KIR2DL4 transfected BaF/3 cells. It does not recognize transfectants that express KIR2DL1, 2DL2, 2DL3, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
181703
Flow Cytometry
Fluorescein
Mouse
Human
Q99706
3805
BaF3 mouse pro-B cell line transfected with human KIR2DL4, Accession # Q99706
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.