Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526053
Description
KIR2DL4 is killer cell immunoglobulin-like receptors (KIRs) which are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC).
Specifications
KIR2DL4 | |
Polyclonal | |
KIR2DL4 | |
CD158D, G9P, KIR103, KIR103AS, CD158 antigen-like family member D, KIR-103AS, MHC class I NK cell receptor KIR103AS, NK cell receptor, killer Ig receptor, killer cell immunoglobulin-like receptor 2DL4, killer cell inhibitory receptor 103AS, natural killer cell inhibitory receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3805 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q99706 | |
KIR2DL4 | |
KLH conjugated synthetic peptide between 296-323 amino acids from the C-terminal region of human KIR2DL4 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction