Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DS4 (CD158i) Mouse anti-Human, Alexa Fluor 647, Clone: 179315, BD

Catalog No. BDB564375 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564375 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564375 Supplier BD Biosciences Supplier No. 564375
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 179315 monoclonal antibody specifically binds to Killer cell immunoglobulin-like receptor 2DS4 which is also known as CD158i, KKA3, and NKAT8. CD158i is encoded by KIR2DS4 (killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4). It is a type I transmembrane glycoprotein that can bind to HLA-C antigens and function as an activating immunoglobulin-like receptor that is expressed on NK cells and some T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DS4 (CD158i)
Applications Flow Cytometry
Classification Monoclonal
Clone 179315
Conjugate Alexa Fluor 647
Description KIR2DS4; KIR antigen 2DS4; KKA3; NKAT8; p58 NK receptor CL-39/CL-1
Host Species Mouse
Immunogen Human KIR2DS4-Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.