Learn More
anti-KIR3DL1, APC, Clone: 177407, R&D Systems™
Description
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | KIR3DL1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 177407 |
| Conjugate | APC |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P43629 |
| Gene Alias | AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, Nkat3, NKAT-3, NKAT3KIR, NKB1, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 |
| Gene Symbols | KIR3DL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.