Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-KIR3DL1, Fluorescein, Clone: 177407, R&D Systems™
Specifications
Antigen | KIR3DL1 |
---|---|
Clone | 177407 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB12251F02
|
R&D Systems
FAB12251F025 |
25 Tests | N/A | N/A | N/A | ||||
FAB12251F10
|
R&D Systems
FAB12251F100 |
100 Tests | N/A | N/A | N/A | ||||
Description
Applications:
Flow Cytometry
Specifications
KIR3DL1 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, Nkat3, NKAT-3, NKAT3KIR, NKB1, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 | |
KIR3DL1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human KIR3DL1 on transfected BaF/3 cells. When tested against a panel of transfectants, no cross-reactivity with KIR2DL1, 2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL2, or 3DS1 is observed. |
177407 | |
Flow Cytometry | |
Fluorescein | |
Mouse | |
Human | |
P43629 | |
3811 | |
BaF3 mouse pro-B cell line transfected with human KIR3DL1, Accession # P43629 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title