Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KiSS1R/GPR54 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216723
Description
KiSS1R/GPR54 Polyclonal specifically detects KiSS1R/GPR54 in Human samples. It is validated for Western Blot.Specifications
KiSS1R/GPR54 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 50% Glycerol with 0.01% Thimerosal | |
AXOR12hypogonadotropin-1, G protein-coupled receptor 54, GPR54kiSS-1R, G-protein coupled receptor 54, G-protein coupled receptor OT7T175, HOT7T175, Hypogonadotropin-1, kiSS-1 receptor, KISS1 receptor, KiSS-1R, Kisspeptins receptor, Metastin receptor | |
Rabbit | |
43 kDa | |
0.1 mL | |
GPCR | |
84634 | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q969F8 | |
KISS1R | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human GPR54. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: mouse (93%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction