Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kruppel-like factor 11, Mouse, Clone: 10D8, Abnova™

Catalog No. 89001082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-082 Supplier Abnova Corporation Supplier No. H00008462M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant KLF11.

Sequence: TYFKSSHLKAHLRTHTGEKPFNCSWDGCDKKFARSDELSRHRRTHTGEKKFVCPVCDRRFMRSDHLTKHARRHMTTKKIPGWQAEVGKLNRIASAESPGSPLVSMPASA

Specifications

Antigen Kruppel-like factor 11
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 10D8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant KLF11.
Formulation PBS with no preservative; pH 7.4
Gene KLF11
Gene Accession No. NM_003597
Gene Alias FKLF/FKLF1/MODY7/TIEG2/Tieg3
Gene Symbols KLF11
Host Species Mouse
Immunogen KLF11 (NP_003588, 404 a.a. ∼ 512 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8462
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.