Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kruppel-like factor 13, Mouse, Clone: 1D3, Abnova™

Catalog No. 89001285 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-285 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-285 Supplier Abnova Corporation Supplier No. H00051621M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant KLF13.

KLF13 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000 [PubMed 11087666]).[supplied by OMIM

Sequence: PESRPEGAAVAATPTLPRVEERRDGKDSASLFVVARILADLN

Specifications

Antigen Kruppel-like factor 13
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant KLF13.
Formulation PBS with no preservative; pH 7.4
Gene KLF13
Gene Accession No. BC010438
Gene Alias BTEB3/FKLF2/NSLP1/RFLAT-1/RFLAT1
Gene Symbols KLF13
Host Species Mouse
Immunogen KLF13 (AAH13946, 31 a.a. ∼ 72 a.a) partial recombinant protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51621
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.