Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klf2 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09820MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-820-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-820-MI Supplier MilliporeSigma Supplier No. 09820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Klf2 Antibody (Rabbit Polyclonal Antibody) validated in western blotting to detect Klf2 also known as Kruppel-like factor 2 (lung), Lung krueppel-like factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Klf2
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Accession No. NP_032478
Host Species Rabbit
Immunogen GST-fusion protein with histidine-tagged mouse Klf2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) Klf2, LKLF
Target Species Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.