Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF5 (Krüppel-like factor 5) Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™

Catalog No. 071580MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-158-0MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-158-0MI Supplier MilliporeSigma Supplier No. 071580

Rabbit Polyclonal Antibody

Use Anti-KLF5 (Krüppel-like factor 5) Antibody (Rabbit Polyclonal Antibody) validated in western blot, Chimmunoprecipitation, immunohistochemistry to detect KLF5 also known as Krüppel-like factor 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF5 (Krüppel-like factor 5)
Applications ChIP Assay, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified rabbit polyclonal serum with 0.05% NaN3
Gene Symbols CKLF ; BTEB2 ; IKLF ; OTTHUMP00000018502
Host Species Rabbit
Immunogen The immunogen was a KLH-conjugated synthetic peptide corresponding to amino acids 95-111 of mouse/rat KLF5.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_033899.2
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial to pellet the solution. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.