Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF9 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535168
Description
This antibody is predicted to react with mouse, porcine and rat based on sequence homology.
The protein encoded by this gene is a transcription factor that binds to GC box elements located in the promoter. Binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription.
Specifications
| KLF9 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q13886 | |
| KLF9 | |
| KLH conjugated synthetic peptide between 29-57 amino acids from the N-terminal region of human KLF9 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| KLF9 | |
| KLF9; BTEB; BTEB1; Krueppel-like factor 9; GC-box-binding protein 1; Basic transcription element-binding protein 1; Transcription factor BTEB1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction