Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
kelch domain containing 4, Mouse, Clone: 4G11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant KLHDC4.
Supplier: Abnova Corporation H00054758M02
Description
Sequence: MGKKGKKEKKGRGAEKTAAKMEKKVSKRSRKEETFLYNELYVYNTRKDTWTKVDIPSPPPRRCAHQAVVVPQGGGQLWVFGGEFASPNGEQFYHYKDLWVLHLATKTWEQVKSTGGPSGRSGHRMVAWKRQLILFGGFHESTRDYIYYNDVYAFNLDTFTWSKLSPSGTGPTPRSGCQMSVTPQGGIVVYGGYSKQRVKKDVDKGTRHSDMFLLKPEDGREDKWVWTRMNPSGVKPTPRSGFSVAMAPNHQTLFFGGVCDEEEEESLSGEFFNDLYFYDATRNRWFEGQLKGPKSEKKKRRRGRKEEPEGGSRPACGGAGTQGPVQLVKEVVAEDGTVVTIKQVLTAPGSAGQPRSEDEDSLEEAGSPAPGPCPRSNAMLAVKHGVLYVYGGMFEAGDRQVTLSDLHCLDLHRMEAWKALVEMDPETQEWLEETDSEEDSEEVEGAEGGVDDEDSGEESGAEDSpecifications
kelch domain containing 4 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC001044 | |
KLHDC4 | |
KLHDC4 (AAH01044, 1 a.a. ∼ 463 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
54758 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
4G11 | |
Mouse monoclonal antibody raised against a full length recombinant KLHDC4. | |
KLHDC4 | |
DKFZp434G0522/FLJ00104 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction