Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

kelch-like 20 (Drosophila), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017057 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-057 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-057 Supplier Abnova Corporation Supplier No. H00027252B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human KLHL20 protein.

The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 44-56 amino acid repeat motif. The kelch motif appears in many different polypeptide contexts and contains multiple potential protein-protein contact sites. Members of this family are present both throughout the cell and extracellularly, with diverse activities. [provided by RefSeq

Sequence: MEGKPMRRCTNIRPGETGMDVTSRCTLGDPNKLPEGVPQPARMPYISDKHPRQTLEVINLLRKHRELCDVVLVVGAKKIYAHRVILSACSPYFRAMFTGELAESRQTEVVIRDIDERAMELLIDFAYTSQITVEEGNVQTLLPAACLLQLAEIQEACCEFLKRQLDPSNCLGIRAFADTHSCRELLRIADKFTQHNFQEVMESEEFMLLPANQLIDIISSDELNVRSEEQVFNAVMAWVKYSIQERRPQLPQVLQHVRLPLLSPKFLVGTVGSDPLIKSDEECRDLVDEAKNYLLLPQERPLMQGPRTRPRKPIRCGEVLFAVGGWCSGDAISSVERYDPQTNEWRMVASMSKRRCGVGVSVLDDLLYAVGGHDGSSYLNSVERYDPKTNQWSSDVAPTSTCRTSVGVAVLGGFLYAVGGQDGVSCLNIVERYDPKENKWTRVASMSTRRLGVAVAVLGGFLYAVGGSDGTSPLNTVERYNPQENRWHTIAPMGTRRKHLGCAVYQDMIYAVGGRDDTTELSSAERYNPRTNQWSPVVAMTSRRSGVGLAVVNGQLMAVGGFDGTTYLKTIEVFDPDANTWRLYGGMNYRRLGGGVGVIKMTHCESHIW

Specifications

Antigen kelch-like 20 (Drosophila)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human KLHL20 protein.
Formulation PBS with no preservative; pH 7.4
Gene KLHL20
Gene Accession No. NM_014458.3
Gene Alias KHLHX/KLEIP/KLHLX/RP3-383J4.3
Gene Symbols KLHL20
Host Species Mouse
Immunogen KLHL20 (NP_055273.2, 1 a.a. ∼ 609 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 27252
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.