Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Klotho, Polyclonal

Catalog No. 42350050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
42-350-050UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-350-050UG Supplier MilliporeSigma™ Supplier No. 42350050UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Klotho Clone: in Mouse samples, and it is validated for ELISA, Immuno Blotting

Inactivation mutation of the recently discoveredklotho gene in mice causes a syndrome resembling aging. Homozygous mutant klotho (kl/kl) mice exhibit premature aging phenotypes such as osteoporosis, age related skin changes, ectopic calcification, atrophy of genital organs and thymus, atherosclerosis, emphysema, ataxia, and short life span. Thekl gene encodes a membrane protein whose extracellular domain carries homology toβ-glucosidases. There also exists a splice variant ofkl mRNA that encodes a putative secreted protein in both human and mouse.kl gene expression was observed to be tissue specific. Improvement of systemic aging phenotypes inkl/kl mice occurs even when the exogenous expression was limited to some organs, suggesting that KL associated aging is regulated through a humoral signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Klotho
Applications ELISA, Immunoblot, Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 0.1% BSA, pH 7.5.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to a 17-amino acid sequence near the C-terminus of murine Klotho
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽100kDa Klotho protein.
Target Species Mouse
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.