Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRG1 Hamster anti-Mouse, APC, Clone: 2F1, BD

Catalog No. BDB561620 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561620 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561620 Supplier BD Biosciences Supplier No. 561620
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 2F1 monoclonal antibody specifically binds to KLRG1 (Killer cell Lectin-like R), which is the mouse homologue of the rat mast cell function-associated antigen (MAFA),on all mouse strains tested (eg, AKR/J, BALB/c, C3H/HeN, C3H.SW, C57BL/6, DBA/1, SJL, 129/J). Unlike rat MAFA, which is expressed on mast cells, mouse KLRG1 is expressed on a large subset of NK cells, lymphokine-activated killer (LAK) cells, adherent LAK (A-LAK) cells, subsets of activated CD8+ T lymphocytes, and small fractions of CD4+ and CD8+ T cells, but not mast cells. The expression of KLRG1 is correlated with reduced proliferative capacity of activated T lymphocytesor reduced effector functions of activated NK cells. This molecule is believed to play a common role in the regulation of leukocytes of both the innate and adaptive immune system. It has been observed that the 2F1 mAb stains the rat basophilic leukemia cell line, RBL-2H3, which is known toexpress MAFA. The KLRG1 protein is an inhibitory lectin-like type IItransmembrane receptor containing a cytoplasmic motif similar to ITIM (Immunoreceptor Tyrosine-based Inhibitory Motif);its ligand has not beenidentified KLRG1 is expressed mainly as a homodimeric molecule consisting of two N-glycosylated subunits of approximately 30-38 kDa. The level of KLRG1 expression is reduced in MHC class I-deficient mice, although direct binding of KLRG1 to MHC class I antigens could not be detected. Cross-linking of KLRG1 by 2F1 mAb reduces TCR-mediated Ca++mobilization and cytotoxic responses (but not IFN-γ production) by CD8+ T cellsand inhibits IFN-γ

TRUSTED_SUSTAINABILITY

Specifications

Antigen KLRG1
Applications Flow Cytometry
Classification Monoclonal
Clone 2F1
Concentration 0.2mg/mL
Conjugate APC
Description Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen A-LAK from C57BL/6 mice
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.