Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRG1 Syrian Hamster anti-Mouse, FITC, Clone: 2F1, MilliporeSigma™

Catalog No. MABF468MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF468MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF468MI Supplier MilliporeSigma Supplier No. MABF468
Add to Cart
Add to Cart

Syrian Hamster Monoclonal Antibody

This golden syrian hamster monoclonal antibody Anti-KLRG1 (mouse), FITC, clone 2F1 validated for use in immunoprecipitation and Flow Cytometry for the detection of KLRG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLRG1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 2F1
Concentration 0.5mg/mL
Conjugate FITC
Formulation Purified Golden Syrian Hamster Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols Klrg1, Mafa
Host Species Syrian Hamster
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.