Learn More
Description
Specifically detects Kpnβ1 Clone: in Human, Mouse, Xenopus samples, and it is validated for Immunoprecipitation
Specifications
Specifications
| Antigen | Kpnβ1 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | In Tris-citrate/phosphate buffer, pH 7-8. |
| Host Species | Rabbit |
| Immunogen | a synthetic peptide corresponding to amino acids near the C-terminus of human Kpnβ1 |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.