Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRAS, Mouse, Clone: 1E4-1B10, Abnova™

Catalog No. 89123205 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-205 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-205 Supplier Abnova Corporation Supplier No. H00003845M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant KRAS.

This gene, a Kirsten ras oncogene homolog from the mammalian ras gene family, encodes a protein that is a member of the small GTPase superfamily. A single amino acid substitution is responsible for an activating mutation. The transforming protein that results is implicated in various malignancies, including lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas and colorectal carcinoma. Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region. [provided by RefSeq

Sequence: MTEYKLVVVGAGGVGKSALTIQLIQNHFVDEYDPTIEDSYRKQVVIDGETCLLDILDTAGHEEYSAMRDQYMRTGEGFLCVFAINNTKSFEDIHHYREQIKRVKDSEDVPMVLVGNKCDLPSRTVDTKQAQDLARSYGIPFIETSAKTRQGVDDAFYTLVREIRKHKEKMSKDGKKKKKKSKTKCVIM

Specifications

Antigen KRAS
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E4-1B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant KRAS.
Formulation PBS with no preservative; pH 7.4
Gene KRAS
Gene Accession No. BC013572
Gene Alias C-K-RAS/K-RAS2A/K-RAS2B/K-RAS4A/K-RAS4B/KI-RAS/KRAS1/KRAS2/NS3/RASK2
Gene Symbols KRAS
Host Species Mouse
Immunogen KRAS (AAH13572, 1 a.a. ∼ 188 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3845
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.