Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

kringle containing transmembrane protein 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004737 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-737 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-737 Supplier Abnova Corporation Supplier No. H00083999A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant KREMEN1.

This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq

Sequence: INQAQGFAVLYQAVKEELPQERPAVNQTVAEVITEQANLSVSAARSSKVLYVITTSPSHPPQTVPGSNSWAPPMGAGSHRVE

Specifications

Antigen kringle containing transmembrane protein 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KREMEN1.
Formulation 50% glycerol
Gene KREMEN1
Gene Accession No. NM_032045
Gene Alias FLJ31863/KREMEM1/KREMEN/KRM1
Gene Symbols KREMEN1
Host Species Mouse
Immunogen KREMEN1 (NP_114434, 310 a.a. ∼ 391 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 83999
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.