Learn More
Description
This gene encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex. [provided by RefSeq
Specifications
Specifications
| Antigen | keratin 17 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 2D10 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant KRT17. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | KRT17 |
| Gene Accession No. | BC000159 |
| Gene Alias | K17/PC/PC2/PCHC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.