Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT80 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524360
Description
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene′s expression profile shows that it encodes a type II epithelial keratin, although structurally the encoded protein is more like a type II hair keratin. This protein is involved in cell differentiation, localizing near desmosomal plaques in earlier stages of differentiation but then dispersing throughout the cytoplasm in terminally differentiating cells. The type II keratins are clustered in a region of chromosome 12q13. Two transcript variants encoding two different fully functional isoforms have been found for this gene.
Specifications
KRT80 | |
Polyclonal | |
Unconjugated | |
Q6KB66 | |
KRT80 | |
KLH conjugated synthetic peptide between 404-432 amino acids from the C-terminal region of human KRT80 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
KRT80 | |
KB20, CK-80, K80, cytokeratin-80, keratin b20, keratin, type II cytoskeletal 80, keratin-80, type II keratin, type-II keratin Kb20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144501 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction