Learn More
Description
Proteins of the Ras superfamily play critical roles in the control of normal and neoplastic proliferation. In mammalian cells there are four true Ras proteins (encoded by Ha-ras, N-ras, Ki-rasA, and Ki-rasB). These proteins relay signals from tyrosine kinases at the plasma membrane to the nucleus via a network of Ser/Thr kinases that includes the MAP kinase (Raf-MEK-ERK) pathway. Kinase suppressor of Ras (KSR-1) was discovered in a genetic screen to identify mutations that suppress constitutively active Ras mutants. KSR-1 contains five domains that are conserved areas (CA) in Drosophila, mouse, and human. CA1 is a unique domain, CA2 is a proline-rich domain, CA3 is a cysteine-rich domain, CA4 is a Ser/Thr rich region, and CA5 contains a C-terminal kinase domain similar to that of Raf. In COS7 cells, ceramide induces the autophosphorylation of KSR-1 and KSR-1-induced phosphorylation and activation of Raf. In addition, overexpression of KSR-1 in 293T cells leads to recruitment of MEK to a 700kDa protein complex and translocates MEK from the soluble to the membrane-associated fraction. Thus, KSR-1 may have important kinase and scaffolding activities during Ras-related signaling.
Host Species: Mouse
Clone: 15
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse KSR-1 aa. 90-203
Formula Weight [Chemical]: 115kDa
Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | KSR-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 15 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Mouse KSR-1 aa. 90-203 |
| Purification Method | Affinity Purified |
| Quantity | 150 μg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.