Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

kinase suppressor of ras 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016637 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-637 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-637 Supplier Abnova Corporation Supplier No. H00283455B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human KSR2 protein.

Sequence: MTDEQVCETVEKYGANREECARLNASLSCLRNVHMSGGNLSKQDWTIQWPTTETGKENNPVCPPEPTPWIRTHLSQSPRVPSKCVQHYCHTSPTPGAPVYTHVDRLTVDAYPGLCPPPPLESGHRSLPPSPRQRHAVRTPPRTPNIVTTVTPPGTPPMRKKNKLKPPGTPPPSSRKLIHLIPGFTALHRSKSHEFQLGHRVDEAHTPKAKKKSKPLNLKIHSSVGSCENIPSQQRSPLLSERSLRSFFVGHAPFLPSTPPVHTEANFSANTLSVPRWSPQIPRRDLGNSIKHRFSTKYWMSQTCTVCGKGMLFGLKCKNCKLKCHNKCTKEAPPCHLLIIHRGDPARLVRTESVPCDINNPLRKPPRYSDLHISQTLPKTNKINKDHIPVPYQPDSSSNPSSTTSSTPSSPAPPLPPSATPPSPLHPSPQCTRQQKNFNLPASHYYKYKQQFIFPDVVPVPETPTRAPQVILHPVTSNPILEGNPLLQIEVEPTSENEEVHDEAEESEDDFEEMNLSLLSARSFPRKASQTSIFLQEWDIPFEQLEIGELIGKGRFGQVYHGRWHGEVAIRLIDIERDNEDQLKAFKREVMAYRQTRHENVVLFMGACMSPPHLAIITSLCKGRTLYSVVRDAKIVLDVNKTRQIAQEIVKGMGYLHAKGILHKDLKSKNVFYDNGKVVITDFGLFSISGVLQAGRREDKLRIQNGWLCHLAPEIIRQLSPDTEEDKLPFSKHSDVFALGTIWYELHAREWPFKTQPAEAIIWQMGTGMKPNLSQIGMGKEISDILLFCWAFEQEERPTFTKLMDMLEKLPKRNRRLSHPGHFWKSAEL

Specifications

Antigen kinase suppressor of ras 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human KSR2 protein.
Formulation PBS with no preservative; pH 7.4
Gene KSR2
Gene Accession No. NM_173598.2
Gene Alias FLJ25965
Gene Symbols KSR2
Host Species Mouse
Immunogen KSR2 (NP_775869.2, 1 a.a. ∼ 829 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 283455
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.