Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KV1.2 (KCNA2) Rabbit anti-Bovine, Canine, Human, Mouse, Porcine, Rabbit, Rat, Xenopus laevis, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523011
Description
Suggested positive control: HEK293 cell lysate.
Kcna2 is a voltage-gated potassium channel (KV) that belongs to the 6-TM family of potassium channel and also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). The Kcna2 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. Kcna2 contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. Further, Kcna2 belongs to the delayed rectifier class, members of which allow nerve cells to efficiently repolarize following an action potential. The coding region of the Kcna2 gene is intronless, and the gene is clustered with genes KCNA3 and KCNA10 on chromosome 1. Diseases associated with KCNA2 include Epileptic Encephalopathy (Early Infantile, 32) and Undetermined Early-Onset Epileptic Encephalopathy.
Specifications
KV1.2 (KCNA2) | |
Polyclonal | |
KCNA2 | |
RP11-284N8.1, HBK5, HK4, HUKIV, KV1.2, MK2, NGK1, RBK2, potassium voltage-gated channel subfamily A member 2, voltage-gated K(+) channel HuKIV, voltage-gated potassium channel HBK5, voltage-gated potassium channel protein Kv1.2, voltage-gated potassium channel subunit Kv1.2 | |
Rabbit | |
100 μL | |
Primary | |
Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat, X. laevis | |
Antibody | |
IgG |
Immunoprecipitation, Neutralization, Western Blot | |
Unconjugated | |
Q28293, P16389, P63141, Q09081, P63142 | |
KCNA2, kcna2.S | |
Synthetic peptide made to a portion of rat Kv1.2 (within residues 325-375). | |
RUO | |
100009479, 16490, 25468, 3737, 373840, 397292, 404022, 539608 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction