Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L1CAM, Mouse, Clone: 3B10, Abnova™

Catalog No. 89123206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-206 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-206 Supplier Abnova Corporation Supplier No. H00003897M11
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant L1CAM.

The protein encoded by this gene is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause three X-linked neurological syndromes known by the acronym CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of a neuron-specific exon is thought to be functionally relevant. [provided by RefSeq

Sequence: PEEYEGHHVMEPPVITEQSPRRLVVFPTDDISLKCEASGKPEVQFRWTRDGVHFKPKEELGVTVYQSPHSGSFTITGNNSNFAQRFQGIYRCFASNKLGTAMSHEIRLMA

Specifications

Antigen L1CAM
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant L1CAM.
Formulation PBS with no preservative; pH 7.4
Gene L1CAM
Gene Accession No. NM_000425
Gene Alias CAML1/CD171/HSAS/HSAS1/MASA/MIC5/N-CAML1/S10/SPG1
Gene Symbols L1CAM
Host Species Mouse
Immunogen L1CAM (NP_000416, 23 a.a. ∼ 132 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3897
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.