Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LacI Mouse anti-E. coli, Clone: 9A5, MilliporeSigma™

Catalog No. 05503IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-503-IMI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-503-IMI Supplier MilliporeSigma Supplier No. 05503I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-LacI Antibody, clone 9A5 is an antibody against LacI for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LacI
Applications Western Blot
Classification Monoclonal
Clone 9A5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide (This antibody is purified from supernatent.).
Gene Accession No. NP_414879
Gene Symbols lacI
Host Species Mouse
Immunogen TrpE fused with full-length E. Coli LacI.
Purification Method Protein G purified
Quantity 200 μL
Primary or Secondary Primary
Target Species E. coli
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.