Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR-1 Mouse anti-Human, Unlabeled, Clone: 14, BD

Catalog No. BDB610974 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610974 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610974 Supplier BD Biosciences Supplier No. 610974

Mouse Monoclonal Antibody

Natural killer (NK) cells are a lymphocyte subpopulation that functions in innate immunity. Unlike cytotoxic T cells, NK cells specifically lyse targets that lack surface MHC class I expression. Inhibitory receptors on the surface of NK cells promote this specificity via the down regulation of lytic activity in response to target cell MHC expression. LAIR-1 (leukocyte-associated immunoglobulin-like receptor-1) is an inhibitory receptor that is consitituitively expressed on human peripheral blood mononuclear leukocytes. The structural characteristics of LAIR-1 include a single extracellular immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory motifs (ITIMs). LAIR-1 crosslinking induces the recruitment of the protein phosphatases SHP-1 and SHP-2 to the ITIMs. These molecules are likely candidates to mediate the negative signal generated by LAIR-1. Although LAIR-1 binding inhibits NK activity, it is not thought to interact with MHC class I. Thus, LAIR-1 is a novel inhibitory receptor that functions independently of MHC recognition to repress the activity of NK cells and possibly, other mononuclear leukocytes. LAIR-1 has been reported to have a calculated molecular weight of 32 kD, but is observable at 40 kD.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAIR-1
Applications Western Blot
Classification Monoclonal
Clone 14
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human LAIR-1 aa. 188-287
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.