Learn More
LAIR-1 Mouse anti-Human, Unlabeled, Clone: DX26, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550810
Description
Reacts with leukocyte associated immunoglobulin-like receptor-1 (LAIR-1), a 32kDa transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory motifs. It is expressed o
Host Species: Mouse
Clone: DX26
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
LAIR-1 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human NK Cell Clone | |
0.1 mg | |
Cell Biology | |
Human | |
IgG1 κ |
Flow Cytometry | |
DX26 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.