Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

laminin, alpha 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017807 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-807 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-807 Supplier Abnova Corporation Supplier No. H00003911D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human LAMA5 protein.

Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined. [provided by RefSeq

Sequence: MGVSLRDKKVHWVYQLGEAGPAVLSIDEDIGEQFAAVSLDRTLQFGHMSVTVERQMIQETKGDTVAPGAEGLLNLRPDDFVFYVGGYPSTFTPPPLLRFPGYRGCIEMDTLNEEVVSLYNFERTFQLDTAVDRPCARSKSTGDPWLTDGSYLDGTGFARISFDSQISTTKRFEQELRLVSYSGVLFFLKQQIQVFLLGGSRKRVLVRVERATVYSVEQDNDLELADAYYLGGVPPDQLPPSLRRLFPTGGSVRGCVKGIKALGKYVDLKRLNTTGVSAGCTADLLVGRAMTFHGHGFLRLALSNVAPLTGNVYSGFGFHSAQDSALLYYRASPDGLCQVSLQQGRVSLQLLRTEVKTQAGFADGAPHYVAFYSNATGVWLYVDDQLQQMKPHRGPPPELQPQPEGPPRLLLGGLPESGTIYNFSGCISNVFVQRLLGPQRVFDLQQNLGSVNVSTGCAPALQAQTPGLGPRGLQATARKASRRSRQPARHPACMLPPHLRTTRDSYQFGGSLSSHLEFVGILARHRNWPSLSMHVLPRSSRGLLLFTARLRPGSPSLALFLSNGHFVAQMEGLGTRLRAQSRQRSRPGRWHKVSVRWEKNRILLVTDGARAWSQEGPHRQHQGAEHPQPHTLFVGGLPASSHSSKLPTSQELHCLMWAWNWRCGPWQSPD

Specifications

Antigen laminin, alpha 5
Applications In situ PLA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human LAMA5 protein.
Formulation PBS with no preservative; pH 7.4
Gene LAMA5
Gene Accession No. BC003355.1
Gene Alias KIAA1907
Gene Symbols LAMA5
Host Species Rabbit
Immunogen LAMA5 (AAH03355.1, 1 a.a. ∼ 670 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 3911
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.