Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LAMB1, Monoclonal, Abnova™

Catalog No. 89208562 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-208-562 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-208-562 Supplier Abnova Corporation Supplier No. H00003912K

Host Species: Rabbit
Isotype:IgG
Species Reactivity:Hu
Immunogen:A synthetic peptide of human LAMB1

Customer may provide cell or tissue lysate for antibody screening.
Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Enzyme Linked Immunosorbent Assay, Western Blotting

Order Info

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Specifications

Antigen LAMB1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description laminin, beta 1
Formulation In 1X PBS, pH 7.4
Gene LAMB1
Gene Accession No. LAMB1
Gene Alias CLM, MGC142015
Gene Symbols LAMB1
Host Species Rabbit
Immunogen A synthetic peptide of human LAMB1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3912
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.