Learn More
Lambda, Ig Light Chain Mouse anti-Human, Unlabeled, Clone: 1-155-2, BD
Shop All BD Biosciences ProductsDescription
Anti-Lambda is specific for lambda light chains of human immunoglobulins. Immunoglobulins bearing lambda light chains are present on approximately 40% of normal B lymphocytes and on Igλ+ neoplastic cells. In serum, Anti-Lambda reacts with immunoglobulins bearing lambda light chains as well as free lambda light chains.
Host Species: Mouse
Clone: 1-55-2
Isotype: IgG1, λ
Cross Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | Lambda, Ig Light Chain |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1-155-2 |
| Concentration | 50μg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human IgA |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.